Learn More
Furfuryl mercaptan, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 119810250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Furfuryl mercaptan | |
98-02-2 | |
1.1300g/mL | |
45°C | |
96% min. (GC) | |
C5H6OS | |
MFCD00003254 | |
17, II, 116 | |
furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol | |
ZFFTZDQKIXPDAF-UHFFFAOYSA-N | |
furan-2-ylmethanethiol | |
7363 | |
97% |
97% | |
Colorless to Yellow | |
155.0°C | |
Authentic | |
Glass bottle | |
1.5290 to 1.5330 | |
25 mL | |
1.13 | |
Solubility in water: insoluble in water. Other solubilities: slightly soluble in chloroform | |
C1=COC(=C1)CS | |
114.16 | |
114.16 | |
Liquid |
Chemical Identifiers
98-02-2 | |
114.16 | |
ZFFTZDQKIXPDAF-UHFFFAOYSA-N | |
7363 | |
C1=COC(=C1)CS |
C5H6OS | |
MFCD00003254 | |
furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol | |
furan-2-ylmethanethiol |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-628-2
RTECSNumber : LU2100000
TSCA : TSCA
RUO – Research Use Only