Please login to your online account to display your discounted pricing

Alfa Aesar™ Furfuryl isopropyl sulfide, 99%

$57.57 - $200.41

Chemical Identifiers

Chemical Name or Material Furfuryl isopropyl sulfide
CAS 1883-78-9
Molecular Formula C8H12OS
Formula Weight 156.25
MDL Number MFCD00040265
Beilstein 1306593
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2199406

  SDS
alfa aesar™
B2199406
5g Each for $57.57

AAB2199414

  SDS
alfa aesar™
B2199414
25g Each for $200.41
Description & Specifications

Specifications

Formula Weight 156.25
Chemical Name or Material Furfuryl isopropyl sulfide
TSCA TSCA
Assay 99%
Molecular Formula C8H12OS
MDL Number MFCD00040265
CAS 1883-78-9
Beilstein 1306593
Health Hazard 1 Warning
Boiling Point 83° to 85°C (16mmHg)
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Quantity 5g
Flash Point 76°C (169°F)
Odor Garlic-like
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.