Please login to your online account to display your discounted pricing

Furfuryl glycidyl ether, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  348490250

 View more versions of this product

Catalog No. AC348490250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0% (GC)
Formula Weight 154.17
Physical Form Liquid
Quantity 25g
Color Colorless to yellow to orange
EINECS Number 226-372-6
RTECS Number LU1423000
Density 1.1220g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May form explosive peroxides.
Refractive Index 1.4800 to 1.4820 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 5380-87-0
Chemical Name or Material Furfuryl glycidyl ether
Assay Percent Range 97%
Molecular Formula C8H10O3
MDL Number MFCD00013348

Specifications

Chemical Name or Material Furfuryl glycidyl ether
CAS 5380-87-0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C8H10O3
Formula Weight 154.17
MDL Number MFCD00013348
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
May form explosive peroxides.

SDS & Safety Documents