missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfuryl Alcohol, TCI America™

Manufacturer:  TCI America F0076500ML

 View more versions of this product

Catalog No. F0076500ML



Specifications

Specifications

Furfuryl Alcohol
Yellow
500mL
C5H6O2
2874
XPFVYQJUAUNWIW-UHFFFAOYSA-N
furan-2-ylmethanol
7361
98.10
≥98.0% (GC)
170°C
-29°C
98-00-0
MFCD00003252
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
C1=COC(=C1)CO
98.101
CHEBI:207496
Clear Liquid at 20°C

Chemical Identifiers

98-00-0
98.101
XPFVYQJUAUNWIW-UHFFFAOYSA-N
7361
furan-2-ylmethanol
C5H6O2
MFCD00003252
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
CHEBI:207496
C1=COC(=C1)CO
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0031

RTECSNumber : LU9100000

TSCA : Yes

Documents