missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfuryl Alcohol, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 119792500

 View more versions of this product

Catalog No. AC119792500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Furfuryl alcohol
-29°C
1.1300g/mL
65°C
97.5% min. (GC)
C5H6O2
MFCD00003252
17, V,3, 338
1.13
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
XPFVYQJUAUNWIW-UHFFFAOYSA-N
furan-2-ylmethanol
7361
CHEBI:207496
98%
98-00-0
Yellow
170°C
Authentic
Glass bottle
1.4850 to 1.488
250 mL
01,408
15, 4334
Solubility in water: miscible but unstable
C1=COC(=C1)CO
98.1
5 mPa.s (25°C)
98.1
Liquid

Chemical Identifiers

98-00-0
98.1
XPFVYQJUAUNWIW-UHFFFAOYSA-N
7361
furan-2-ylmethanol
C5H6O2
MFCD00003252
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
CHEBI:207496
C1=COC(=C1)CO
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer if inhaled.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Toxic if swallowed.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Imm

GHS Signal Word: Danger

EINECSNumber : 202-626-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only