missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Furfuryl acetate, 97%

Manufacturer:  Alfa Aesar™ A1528536

 View more versions of this product

Catalog No. AAA1528536



Description

Description

Applications
Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Furfuryl acetate
175°C to 177°C
623-17-6
MFCD00003251
furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
CC(=O)OCC1=CC=CO1
140.138
140.14
65°C (149°F)
500g
C7H8O3
116128
CKOYRRWBOKMNRG-UHFFFAOYSA-N
furan-2-ylmethyl acetate
12170
97%

Chemical Identifiers

623-17-6
140.138
CKOYRRWBOKMNRG-UHFFFAOYSA-N
12170
CC(=O)OCC1=CC=CO1
C7H8O3
MFCD00003251
furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
furan-2-ylmethyl acetate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LU9120000

TSCA : TSCA

Documents