Please login to your online account to display your discounted pricing

Furegrelate Sodium Salt, ≥99%

Inhibitor of thrombaxane synthetase

Specifications

Assay ≥99%
CAS 85666-17-7
Formula Weight 275.20g/mol
Synonym U-63557A; Sodium furegrelate
Storage Requirements −30° to −10°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ61378LB0

 
alfa aesar™
J61378LB0
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99%
CAS 85666-17-7
Formula Weight 275.20g/mol
Synonym U-63557A; Sodium furegrelate
Storage Requirements −30° to −10°C
Format Crystalline Solid
For Use With (Application) Inhibitor of thrombaxane synthetase
Description White to Off-white
Solubility Soluble in water at approximately 15mg/ml