missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furan (stabilized with BHT), TCI America™

Manufacturer:  TCI America F007425ML

 View more versions of this product

Catalog No. F007425ML



Specifications

Specifications

Furan (stabilized with BHT)
Yellow
25mL
C4H4O
2389
YLQBMQCUIZJEEH-UHFFFAOYSA-N
furan
8029
68.08
≥99.0% (GC)
31°C
-84°C
110-00-9
MFCD00003222
divinylene oxide, furfuran, tetrole, oxole, oxacyclopentadiene, furane, 1,4-epoxy-1,3-butadiene, axole, rcra waste number u124, furfurane
C1=COC=C1
68.075
CHEBI:35559
Clear Liquid at 20°C

Chemical Identifiers

110-00-9
68.075
YLQBMQCUIZJEEH-UHFFFAOYSA-N
8029
furan
C4H4O
MFCD00003222
divinylene oxide, furfuran, tetrole, oxole, oxacyclopentadiene, furane, 1,4-epoxy-1,3-butadiene, axole, rcra waste number u124, furfurane
CHEBI:35559
C1=COC=C1
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3334

RTECSNumber : LT8524000

TSCA : Yes

Documents