Learn More
Furan-3-sulfonyl chloride, ≥97%, Thermo Scientific™
$282.84 - $282.84
Chemical Identifiers
CAS | 52665-49-3 |
---|---|
Molecular Formula | C4H3ClO3S |
Molecular Weight (g/mol) | 166.575 |
MDL Number | MFCD08271875 |
InChI Key | PCBBEIXTVWWTAK-UHFFFAOYSA-N |
Synonym | 3-furansulfonyl chloride, furan-3-sulfonylchloride, chloro-3-furylsulfone, 3-furansulfonylchlorid, 3-furansulfonylchloride, 3-furylsulfonyl chloride, 3-chlorosulphonyl furan, 3-furansulfonyl chloride;, 3-furanylsulfonyl chloride, 3-furylsulfonyl chloride; |
PubChem CID | 22179302 |
IUPAC Name | furan-3-sulfonyl chloride |
SMILES | C1=COC=C1S(=O)(=O)Cl |
Chemical Identifiers
52665-49-3 | |
166.575 | |
PCBBEIXTVWWTAK-UHFFFAOYSA-N | |
22179302 | |
C1=COC=C1S(=O)(=O)Cl |
C4H3ClO3S | |
MFCD08271875 | |
3-furansulfonyl chloride, furan-3-sulfonylchloride, chloro-3-furylsulfone, 3-furansulfonylchlorid, 3-furansulfonylchloride, 3-furylsulfonyl chloride, 3-chlorosulphonyl furan, 3-furansulfonyl chloride;, 3-furanylsulfonyl chloride, 3-furylsulfonyl chloride; | |
furan-3-sulfonyl chloride |
Specifications
52665-49-3 | |
100.0 | |
Amber Glass Bottle | |
MFCD08271875 | |
3-furansulfonyl chloride, furan-3-sulfonylchloride, chloro-3-furylsulfone, 3-furansulfonylchlorid, 3-furansulfonylchloride, 3-furylsulfonyl chloride, 3-chlorosulphonyl furan, 3-furansulfonyl chloride;, 3-furanylsulfonyl chloride, 3-furylsulfonyl chloride; | |
C1=COC=C1S(=O)(=O)Cl | |
166.575 | |
166.58 | |
Furan-3-sulfonyl chloride |
96.5 | |
97% | |
C4H3ClO3S | |
250 mg | |
PCBBEIXTVWWTAK-UHFFFAOYSA-N | |
furan-3-sulfonyl chloride | |
22179302 | |
97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger