Please login to your online account to display your discounted pricing

Alfa Aesar™ Furan-3-boronic acid, 97%

$94.66 - $404.82

Chemical Identifiers

Chemical Name or Material Furan-3-boronic acid
CAS 55552-70-0
Molecular Formula C4H5BO3
Formula Weight 111.89
MDL Number MFCD01319007
Beilstein 1635653
Synonym 3-Furylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1983403

 
alfa aesar™
L1983403
1g Each for $94.66

AAL1983406

 
alfa aesar™
L1983406
5g Each for $404.82
Description & Specifications

Specifications

Formula Weight 111.89
Beilstein 1635653
Melting Point 139° to 144°C
Chemical Name or Material Furan-3-boronic acid
Assay 97%
Molecular Formula C4H5BO3
MDL Number MFCD01319007
CAS 55552-70-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Furylboronic acid
Health Hazard 1 Warning
Quantity 1g

Applications
Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.