Please login to your online account to display your discounted pricing

Alfa Aesar™ Furan-2-thiocarboxamide, 97%

$110.82 - $378.66

Chemical Identifiers

Chemical Name or Material Furan-2-thiocarboxamide
CAS 17572-09-7
Molecular Formula C5H5NOS
Formula Weight 127.16
MDL Number MFCD01935200
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5183103

  SDS
alfa aesar™
H5183103
1g Each for $378.66

AAH51831MD

  SDS
alfa aesar™
H51831MD
250mg Each for $110.82
Description & Specifications

Specifications

Formula Weight 127.16
Chemical Name or Material Furan-2-thiocarboxamide
Assay 97%
Molecular Formula C5H5NOS
MDL Number MFCD01935200
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 17572-09-7
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.