missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furan-2-sulfonyl chloride, ≥97%, Maybridge

$239.94 - $683.59

Chemical Identifiers

CAS 52665-48-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C4H3ClO3S
Formula Weight 166.58
MDL Number MFCD06658968
Chemical Name or Material Furan-2-sulfonyl chloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC02003CB SDS Fisher Scientific Company
CC02003CB
Amber Glass Bottle 250mg Each for $239.94
CC02003DA SDS Fisher Scientific Company
CC02003DA
Amber Glass Bottle 1g Each for $683.59
Specifications Description & Specifications

Chemical Identifiers

52665-48-2
100.0
C4H3ClO3S
MFCD06658968
97%
96.5
97%
166.58
Furan-2-sulfonyl chloride

Specifications

52665-48-2
100.0
Amber Glass Bottle
166.58
250mg
97%
96.5
97%
C4H3ClO3S
MFCD06658968
Furan-2-sulfonyl chloride

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger