missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Furan-2-boronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ L1836603

 View more versions of this product

Catalog No. AAL1836603



Specifications

Specifications

Furan-2-boronic acid pinacol ester
1g
C10H15BO3
9045028
GWEWQKZABZXLJH-UHFFFAOYSA-N
2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
11356013
98%
1.472
374790-93-9
MFCD02094008
1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl, 2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furanboronic acid pinacol ester, furan-2-boronic acid pinacol cyclic ester, furan-2-boronic acid pinacol ester, pubchem18440, zlchem 693
B1(OC(C(O1)(C)C)(C)C)C2=CC=CO2
194.037
194.04

Chemical Identifiers

374790-93-9
194.037
GWEWQKZABZXLJH-UHFFFAOYSA-N
11356013
B1(OC(C(O1)(C)C)(C)C)C2=CC=CO2
C10H15BO3
MFCD02094008
1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl, 2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan, 2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-furanboronic acid pinacol ester, furan-2-boronic acid pinacol cyclic ester, furan-2-boronic acid pinacol ester, pubchem18440, zlchem 693
2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents