Please login to your online account to display your discounted pricing

Alfa Aesar™ FURA 2 pentasodium salt

A very sensitive fluorescent calcium chelator

Chemical Identifiers

Chemical Name or Material FURA 2 pentasodium salt
CAS 113694-64-7
Formula Weight 751.45
MDL Number MFCD00083328
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63686MCR

 
alfa aesar™
J63686MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 751.45
Chemical Name or Material FURA 2 pentasodium salt
CAS 113694-64-7
Physical Form Solid
Quantity 1mg
Sensitivity Moisture & Light Sensitive
MDL Number MFCD00083328
Color Yellow
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C