Please login to your online account to display your discounted pricing

Alfa Aesar™ FURA 2-AM

Chemical Identifiers

Chemical Name or Material FURA 2-AM
CAS 108964-32-5
Formula Weight 1001.87
MDL Number MFCD00036976
Beilstein 8183748
Synonym Fura-2 pentakis(acetoxymethyl) ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ62728MCR

 
alfa aesar™
J62728MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1001.87
MDL Number MFCD00036976
CAS 108964-32-5
Chemical Name or Material FURA 2-AM
Quantity 1mg
Physical Form Powder
Beilstein 8183748
Synonym Fura-2 pentakis(acetoxymethyl) ester
Color Yellow
Sensitivity Light Sensitive
Storage Note 1 -30° to -10°C

  • A calcium indicator; derivative of Fura-2 used to load the chelator into cells

Safety and Handling

-30° to -10°C