LaMotte™ Funky Faucet Sink Test Kit
Discover a fascinating world of organisms in tap water.
Manufacturer:  LaMotte™ 5914
Catalog No. S66227
Includes: Nonhazardous TesTabs™ reagents for 10 tests, test tubes, word search game and activities and instructions for simple water quality tests
Description
Description
Ideal teacher demos for elementary students.
  • Test tap water for pH, chlorine, hardness, copper and iron
  • “Just for Fun” activities include “Make Your Own Lava Lamp” and edible experiments like the “Pollution Parfait”
Order Info
Grades 3+.
Specifications
Specifications
Test tap water for pH, chlorine, hardness, copper and iron
3+
Nonhazardous testabs™ reagents for 10 tests, test tubes, word search game and activities and instructions
Funky Faucet Sink Test Kit