Please login to your online account to display your discounted pricing
Fungilab Viscolead Adv Viscometer
Measure dynamic viscosity and kinematics viscosity.
$5,682.23 - $6,845.34
Specifications
Alarm Types Auto-test with sound and visual malfunction alarm
Certifications/Compliance BS: 6075, 5350, ISO: 2555, 1652, ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716
Count (Speed Setting) 18
Repeatability 0.2%
Temperature Range (Metric) 0° to 100°C
View More Specs
Includes:  Standard spindles (4 spindles for L model, and 6 spindles for R and H models), viscometer stand and spindle protector, carrying case, USB cable and Datalogger Software (For data transfer to a PC Excel format)
Products
Catalog Number Mfr. No. Includes Product Type Viscosity Price Quantity    
06-815-019
fungilab
VL201001
PPR spindle coupling system, viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv H Rotational Viscometer 200 to 106.000.000 cP Each for $6,325.83
06-815-022
fungilab
VL211001
PT100 probe, PPR spindle coupling system, viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv H Rotational Viscometer 200 to 106.000.000 cP Each for $6,732.81
06-815-021
fungilab
VL201003
PPR spindle coupling system, viscometer stand and spindle protector, carrying case, 4 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv L Rotational Viscometer 20 to 2.000.000 cP Each for $6,325.83
06-815-023
fungilab
VL211002
PT100 probe, PPR spindle coupling system, viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv R Rotational Viscometer 100 to 40.000.000 cP Each for $6,732.81
06-815-020
fungilab
VL201002
PPR spindle coupling system, viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv R Rotational Viscometer 100 to 13.000.000 cP Each for $6,325.83
06-815-024
fungilab
VL211003
PT100 probe, PPR spindle coupling system, viscometer stand and spindle protector, carrying case, 4 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv L Rotational Viscometer 20 to 6.000.000 cP Each for $6,732.81
06-815-004
fungilab
VL200001
Viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv H Rotational Viscometer 200 to 106.000.000 cP Each for $6,845.34
06-815-006
fungilab
VL200003
Viscometer stand and spindle protector, carrying case, 4 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv R Rotational Viscometer 20 to 2.000.000 cP Each for $5,682.23
06-815-005
fungilab
VL200002
Viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv L Rotational Viscometer 100 to 13.000.000 cP Each for $5,682.23
06-815-007
fungilab
VL210001
PT100 probe, viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv H Rotational Viscometer 200 to 106.000.000 cP Each for $6,089.21
06-815-009
fungilab
VL210003
PT100 probe, viscometer stand and spindle protector, carrying case, 4 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv L Rotational Viscometer 20 to 6.000.000 cP Each for $6,379.47
06-815-008
fungilab
VL210002
PT100 probe, viscometer stand and spindle protector, carrying case, 6 standard spindles, USB cable and Datalogger Software (For data transfer to a PC Excel format) Viscolead Adv R Rotational Viscometer 100 to 40.000.000 cP Each for $6,379.47
Description & Specifications
Specifications
Alarm Types Auto-test with sound and visual malfunction alarm
Certifications/Compliance BS: 6075, 5350, ISO: 2555, 1652, ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716
Count (Speed Setting) 18
Repeatability 0.2%
Temperature Range (Metric) 0° to 100°C
For Use With (Application) Dynamic viscosity and kinematics viscosity
Hertz 50/60Hz
Probe Type PT100
Software Type USB, datalogger software
Speed Range 0.3 to 100rpm
Digital display of selected speed (rpm), selected spindle (SP), viscosity reading in (cP/mPaS/cSt), percentage of full scale (%), sample temperature (°C or °F, Optional), Shear Rate with coaxial spindles SR (s-1), Shear Stress with coaxial spindles SS (N/m2), and user-introduced density (g/cm3).
  • Direct readout
  • SI to CGS Unit converter
  • Time to torque: target torque pre-setting device
  • Time to stop: target time pre-setting device
  • AUTO-TEST with sound and visual malfunction alarm
  • AUTO-RANGE function
  • 10 working memories
  • Option of 10 languages
  • Temperature reading by PT100 (optional)
  • User-enabled viscosity and temperature (optional) calibration
  • AISI 316 stainless steel spindles, identifiable by number and letter
Also available with the Push Plug and Release (PPR) System for spindle coupling.
Conforms to standards:
BS: 6075, 5350
ISO: 2555, 1652
ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716