Fungilab Ubbelohde ASTM and DIN Capillary Viscometers
Manufacturer:  Fungilab CV003103
 View more versions of this product
Catalog No. 06-815-146
Specifications Description & Specifications
Specifications
0.55 lb.
15mL
0.5 oz.
±0.2%
I
0.25kg
Calibration certificate
Ubbelohde Din Viscometer
Yes
DIN 51562 Part 1, ISO 3105, UNE 400313, calibration certificate
Used to determine kinematic viscosity of transparent Newtonian liquids
I
0.01
Glass
2 to 10 cSt