Please login to your online account to display your discounted pricing
Fungilab Spindles for Small Sample Adapters
Stainless steel disc measuring system for relative viscosity measurements in a 600mL beaker.
$237.70 - $1,529.68
Specifications
Certifications/Compliance ASTM, ISO 2555
For Use With (Application) For relative viscosity measurement
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Product Type Price Quantity    
06-815-036
fungilab
V12600N
Small sample adapter Tl5, Tl6, Tl7 Spindle Each for $987.32
06-815-037
fungilab
V12700N
Small sample adapter Tr8, Tr9, Tr10, Tr11 Spindle Each for $1,529.68
06-815-040
fungilab
V12401N
Small sample adapter L1 Spindle Each for $273.70
06-815-039
fungilab
V12400N
Small sample adapter L1, L2, L3, L4 Spindle Each for $825.57
06-815-041
fungilab
V12402N
Small sample adapter L2 Spindle Each for $273.70
06-815-043
fungilab
V12404N
Small sample adapter L4 Spindle Each for $273.70
06-815-044
fungilab
V12500N
Small sample adapter R2, R3, R4, R5, R6, R7 Spindle Each for $1,184.77
06-815-042
fungilab
V12403N
Small sample adapter L3 Spindle Each for $273.70
06-815-045
fungilab
V12501N
Small sample adapter R1 Spindle Each for $412.23
06-815-047
fungilab
V12503N
Small sample adapter R3 Spindle Each for $237.70
06-815-046
fungilab
V12502N
Small sample adapter R2 Spindle Each for $237.70
06-815-048
fungilab
V12504N
Small sample adapter R4 Spindle Each for $237.70
06-815-049
fungilab
V12505N
Small sample adapter R5 Spindle Each for $237.70
06-815-051
fungilab
V12507N
Small sample adapter R7 Spindle Each for $237.70
06-815-050
fungilab
V12506N
Small sample adapter R6 Spindle Each for $237.70
06-815-052
fungilab
V12605N
Small sample adapter Tl5 Spindle Each for $419.38
06-815-053
fungilab
V12606N
Small sample adapter Tl6 Spindle Each for $419.38
06-815-054
fungilab
V12607N
Small sample adapter Tl7 Spindle Each for $419.38
06-815-055
fungilab
V12708N
Small sample adapter Tr8 Spindle Each for $409.79
06-815-056
fungilab
V12709N
Small sample adapter Tr9 Spindle Each for $409.79
06-815-057
fungilab
V12710N
Small sample adapter Tr10 Spindle Each for $409.79
06-815-058
fungilab
V12711N
Small sample adapter Tr11 Spindle Each for $409.79
06-815-059
fungilab
V12800N
Small sample adapter and LCP Spindle Each for $409.79
Description & Specifications
Specifications
Certifications/Compliance ASTM, ISO 2555
For Use With (Application) For relative viscosity measurement
Material Stainless Steel
Spindles for use with Small Sample Adapters (Cat. Nos. 06-815-032, -031).
  • This system is used for measurements in accordance with the ASTM-ISO 2555 standard
  • Standard spindles (included in standard delivery): R and H spindles, L spindles
  • Special spindles (additionally ordered with small sample adapters APM and APM/B): TL and TR spindles
  • Spindle set L: 20 to 6,000,000 cP (for ADV and PRO models); 20 to 2,000,000 cP (for ONE model)
  • Spindle set R: 100 to 40,000,000cP
  • Spindles set R for H models: 200 to 106,000,000cP