Please login to your online account to display your discounted pricing

Fungilab Spindles for Small Sample Adapters


Stainless steel disc measuring system for relative viscosity measurements in a 600mL beaker.

Manufacturer: fungilab  V12605N

 View more versions of this product

Catalog No. 06-815-052

  • / Each
 

Description & Specifications

Specifications

Certifications/Compliance ASTM, ISO 2555
Viscosity 3 to 10,000 cP
Product Type Tl5 Spindle
Material Stainless Steel
For Use With (Application) For relative viscosity measurement
For Use With (Equipment) Small sample adapter

Spindles for use with Small Sample Adapters (Cat. Nos. 06-815-032, -031).

  • This system is used for measurements in accordance with the ASTM-ISO 2555 standard
  • Standard spindles (included in standard delivery): R and H spindles, L spindles
  • Special spindles (additionally ordered with small sample adapters APM and APM/B): TL and TR spindles
  • Spindle set L: 20 to 6,000,000 cP (for ADV and PRO models); 20 to 2,000,000 cP (for ONE model)
  • Spindle set R: 100 to 40,000,000cP
  • Spindles set R for H models: 200 to 106,000,000cP