Please login to your online account to display your discounted pricing

Fungilab Silicone Standard Oils

Silicone standard oils to check and calibrate rotational viscometers

$372.00 - $409.49

Specifications

Certifications/Compliance BS EN ISO / IEC 17025
Product Type Silicone Standard Oil
Volume (Metric) 600mL
For Use With (Application) Used for viscosity tests and meter calibration
For Use With (Equipment) Rotational viscometer
View More Specs
Products
Catalog Number Mfr. No. Viscosity Price Quantity    

06-815-064

 
fungilab
V12Z10
4.972 MPa at 25°C Each for $384.71

06-815-066

 
fungilab
V12Z18
50.33 MPa at 25°C Each for $372.00

06-815-065

 
fungilab
V12Z14
9.972 MPa at 25°C Each for $405.66

06-815-068

 
fungilab
V12Z22
101.66 MPa at 25°C Each for $384.71

06-815-067

 
fungilab
V12Z20
76.33 MPa at 25°C Each for $384.71

06-815-069

 
fungilab
V12Z23
244.1 MPa at 25°C Each for $392.26

06-815-070

 
fungilab
V12Z24
344.1 MPa at 25°C Each for $384.71

06-815-071

 
fungilab
V12Z26
501.9 MPa at 25°C Each for $409.49

06-815-072

 
fungilab
V12Z30
1,019 MPa at 25°C Each for $405.66

06-815-073

 
fungilab
V12Z34
5,094 MPa at 25°C Each for $405.66

06-815-074

 
fungilab
V12Z36
10,061 MPa at 25°C Each for $384.71

06-815-075

 
fungilab
V12Z38
12,309 MPa at 25°C Each for $405.66

06-815-077

 
fungilab
V12Z46
60,586 MPa at 25°C Each for $384.71

06-815-076

 
fungilab
V12Z42
30,240 MPa at 25°C Each for $405.66
Description & Specifications

Specifications

Certifications/Compliance BS EN ISO / IEC 17025
Product Type Silicone Standard Oil
Volume (Metric) 600mL
For Use With (Application) Used for viscosity tests and meter calibration
For Use With (Equipment) Rotational viscometer
Packaging Ready-to-use Packs
Quantity Each
Shelf Life 2 Years
Volume (English) 20.28 oz.
Weight (English) 4.4 lb.
Weight (Metric) 2kg

Silicone viscosity standards are specifically engineered for use with rotational viscometers for viscosity tests and meter calibration.

  • Supplied in 600mL ready-to-use packs making sample transfer to test beaker unnecessary
  • Viscosity values read at 20°C and 25°C temperatures
  • All standard oils are supplied with their own calibration certificate
  • Immediate stock delivery
  • Lowest uncertainty margin
  • Fully comply with international test method protocol
  • Full traceability to national standards
  • Tamper-evident security packaging
  • Two-year shelf life

Certified to International Standard BS EN ISO / IEC 17025.