Fungilab™ Cannon-Fenske Routine Capillary Viscometers
$347.45 - $347.45
Specifications
Calibrated Yes
Certifications/Compliance ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
For Use With (Application) Measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries
Product Type Cannon™ Fenske Routine Viscometer
Includes Calibration certificate
View More Specs
Products 3
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Price Quantity  
Catalog Number Mfr. No. Approximate Constant ASTM Numbers Size Viscosity Price Quantity  
06-815-117 View Documents Fungilab™
CV004203
0.008 75 75 1.6 to 8 cSt Each for $347.45
06-815-122 View Documents Fungilab™
CV004208
0.5 350 350 100 to 500 cSt Each for $347.45
06-815-115 View Documents Fungilab™
CV004201
0.002 25 25 0.5 to 2 cSt Each for $347.45
Specifications
Specifications
Yes
Measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries
Calibration certificate
±0.2%
0.23 oz.
0.25kg
ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
Cannon™ Fenske Routine Viscometer
Glass
7mL
0.55 lb.