missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fungilab™ Cannon-Fenske Opaque Capillary Viscometers

Fungilab Cannon-Fenske Opaque Capillary Viscometers are used for dark Newtonian liquids and to study shearing stress and shearing rate.

$347.45 - $493.63

Specifications

Calibrated Yes
Certifications/Compliance ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
For Use With (Application) Used for dark Newtonian liquids and to study shearing stress and shearing rate
Product Type Cannon™ Fenske Opaque Reverse Flow Viscometer
Includes Calibration certificate
View More Specs

Products 4
Catalog Number Mfr. No. Size Viscosity Price Quantity  
Catalog Number Mfr. No. Size Viscosity Price Quantity  
06-815-107 View Documents Fungilab™
CV004105
150 7 to 35 cSt Each for $347.45
 
06-815-110 View Documents Fungilab™
CV004108
350 100 to 500 cSt Each for $493.63
 
06-815-112 View Documents Fungilab™
CV004110
450 500 to 2500 cSt Each for $388.49
 
06-815-114 View Documents Fungilab™
CV004112
600 4.000 to 20.000 cSt Each for $466.57
 
Description

Description

  • Reverse flow type
  • Constructed of glass
  • Sample Volume: 0.4 oz. (12mL)

Certifications

ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate

Specifications

Specifications

Yes
Used for dark Newtonian liquids and to study shearing stress and shearing rate
Calibration certificate
±0.3%
0.4 oz.
0.25kg
ASTM D445 446, ASTM D2515, ISO 3104 3105, UNE 400313, calibration certificate
Cannon™ Fenske Opaque Reverse Flow Viscometer
Glass
12mL
0.55 lb.