missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fumaronitrile, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 157400250
Specifications
Fumaronitrile | |
Glass bottle | |
White | |
25g | |
97.5% min. (GC) | |
NCCH=CHCN | |
02, I, 302 | |
KYPOHTVBFVELTG-OWOJBTEDSA-N | |
(E)-but-2-enedinitrile | |
637930 | |
Crystals |
Authentic | |
186.0°C | |
95.0°C to 98.0°C | |
764-42-1 | |
C4H2N2 | |
MFCD00001928 | |
2-butenedinitrile, 2e, 2-butenedinitrile, e, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene, fumaric nitrile, fumarodinitrile, fumaronitrile, furmaronitrile, trans-1,2-dicyanoethene, trans-1,2-dicyanoethylene | |
C(=CC#N)C#N | |
78.06 | |
78.06 | |
98% |
Chemical Identifiers
764-42-1 | |
78.06 | |
KYPOHTVBFVELTG-OWOJBTEDSA-N | |
637930 | |
C(=CC#N)C#N |
C4H2N2 | |
MFCD00001928 | |
2-butenedinitrile, 2e, 2-butenedinitrile, e, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene, fumaric nitrile, fumarodinitrile, fumaronitrile, furmaronitrile, trans-1,2-dicyanoethene, trans-1,2-dicyanoethylene | |
(E)-but-2-enedinitrile |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
GHS P Statement
Do not eat,drink or smoke when using this product.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis
GHS Signal Word: Danger
EINECSNumber : 212-122-3