missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fumaric Acid, TCI America™

Manufacturer:  TCI America F006725G

 View more versions of this product

Catalog No. F006725G



Specifications

Specifications

Fumaric Acid
287°C
2.2 to 2.6 (1% H2O suspention)
C4H4O4
fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
C(=CC(=O)O)C(=O)O
116.072
CHEBI:18012
Crystal-Powder at 20°C
White
25g
110-17-8
MFCD00002700
VZCYOOQTPOCHFL-OWOJBTEDSA-N
(E)-but-2-enedioic acid
444972
116.07
≥99.0% (GC,T)

Chemical Identifiers

110-17-8
116.072
VZCYOOQTPOCHFL-OWOJBTEDSA-N
444972
(E)-but-2-enedioic acid
C4H4O4
MFCD00002700
fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
CHEBI:18012
C(=CC(=O)O)C(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1091

RTECSNumber : LS9625000

TSCA : Yes

Documents