Learn More
Fumaric acid, 99+%, ACROS Organics™
Manufacturer: Fisher Scientific Acros AC119751000
Specifications
Fumaric acid | |
230°C | |
Plastic bottle | |
0.25% max. (K.F.) | |
295.0°C to 300.0°C | |
110-17-8 | |
C4H4O4 | |
MFCD00002700 | |
05,319 | |
2-butenedioic acid, 2e-but-2-enedioic acid, allomaleic acid, boletic acid, butenedioic acid, fumaric acid, lichenic acid, trans-2-butenedioic acid, trans-butenedioic acid, tumaric acid | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
(E)-but-2-enedioic acid | |
444972 | |
116.07 | |
99+% |
1.6200g/mL | |
Authentic | |
1.62 | |
White | |
100g | |
99+% | |
HO2CCH=CHCO2H | |
02, IV, 2202 | |
15, 4316 | |
Solubility in water: 6.3g/l (25°C). Other solubilities: 0.72g/100g ether (25°C)- 1.72g/100g acetone,(30°C)- 5.76g/100 g 95% alcohol (30°C),practically insoluble in chloroform,carbon tetra-,chloride and benzene | |
C(=CC(=O)O)C(=O)O | |
116.07 | |
CHEBI:18012 | |
Fine Crystalline Powder |
Chemical Identifiers
110-17-8 | |
116.07 | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
444972 | |
(E)-but-2-enedioic acid |
C4H4O4 | |
MFCD00002700 | |
2-butenedioic acid, 2e-but-2-enedioic acid, allomaleic acid, boletic acid, butenedioic acid, fumaric acid, lichenic acid, trans-2-butenedioic acid, trans-butenedioic acid, tumaric acid | |
CHEBI:18012 | |
C(=CC(=O)O)C(=O)O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-743-