Please login to your online account to display your discounted pricing

Fumaric acid, 99.5%, Acros Organics


Quantity 500g

Manufacturer: thermo fisher scientific  220445000

 View more versions of this product

Catalog No. AC220445000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Physical Form Fine Crystalline Powder
Formula Weight 116.07
Quantity 500g
Color White
EINECS Number 203-743-0
RTECS Number LS9625000
TSCA TSCA
Density 1.62g/mL
Beilstein 02, IV, 2202
Fieser 05,319
Merck Index 15, 4316
Iron (Fe) 5ppm max.
Additional Information total sulfur (S): < 0.0005%
Heavy Metals (as Pb) 5ppm max.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chlorine (Cl) 50ppm max.
Residue after Ignition 0.005% max.
Infrared Spectrum Authentic
Arsenic (As) 2ppm max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Chemical Name or Material Fumaric acid
Molecular Formula C4H4O4
Name Note p.a.
Assay Percent Range 99.50%
CAS 110-17-8
Linear Formula HO2CCH=CHCO2H

Specifications

Chemical Name or Material Fumaric acid
Name Note p.a.
CAS 110-17-8
Assay ≥99.5%
Assay Percent Range 99.50%
Molecular Formula C4H4O4
Linear Formula HO2CCH=CHCO2H
Formula Weight 116.07
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents