missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fumaramide, TCI America™

Manufacturer:  TCI America F006610G

 View more versions of this product

Catalog No. F006610G



Specifications

Specifications

Fumaramide
10g
C4H6N2O2
fumaramide, fumaric diamide, 2-butenediamide, e, 2-butenediamide e, but-2-enediamide, e-but-2-enediamide, 2-butenediamide, 2e, 1e ethene-1,2-dicarboxamide, fumaramid, e-2-butenediamide
C(=CC(=O)N)C(=O)N
114.104
114.10
≥96.0% (N)
White
627-64-5
MFCD00025527
BSSNZUFKXJJCBG-OWOJBTEDSA-N
(E)-but-2-enediamide
446408
Crystal-Powder at 20°C

Chemical Identifiers

627-64-5
114.104
BSSNZUFKXJJCBG-OWOJBTEDSA-N
446408
C(=CC(=O)N)C(=O)N
C4H6N2O2
MFCD00025527
fumaramide, fumaric diamide, 2-butenediamide, e, 2-butenediamide e, but-2-enediamide, e-but-2-enediamide, 2-butenediamide, 2e, 1e ethene-1,2-dicarboxamide, fumaramid, e-2-butenediamide
(E)-but-2-enediamide
Safety and Handling

Safety and Handling

TSCA : No

Documents