Please login to your online account to display your discounted pricing

Fumaramide, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  338300050

 View more versions of this product

Catalog No. AC338300050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 114.1
Quantity 5g
Color White
EINECS Number 211-007-5
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula HNC(O)CHCHC(O)nH
Chemical Name or Material Fumaramide
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4H6N2O2
MDL Number MFCD00025527
CAS 627-64-5

Specifications

Chemical Name or Material Fumaramide
Name Note 98%
CAS 627-64-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H6N2O2
Linear Formula HNC(O)CHCHC(O)nH
Formula Weight 114.1
MDL Number MFCD00025527