Please login to your online account to display your discounted pricing

Fumaramide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Fumaramide
Name Note 98%
CAS 627-64-5
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C4H6N2O2
Linear Formula HNC(O)CHCHC(O)nH
Formula Weight 114.1g/mol
MDL Number MFCD00025527
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC338300050

  SDS
thermo fisher scientific
338300050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Quantity 5g
Chemical Name or Material Fumaramide
Name Note 98%
CAS 627-64-5
Physical Form White powder
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C4H6N2O2
Linear Formula HNC(O)CHCHC(O)nH
Formula Weight 114.1g/mol
MDL Number MFCD00025527
EINECS Number 211-007-5