Please login to your online account to display your discounted pricing

Alfa Aesar™ Fumaramide, 97%

$64.15 - $529.65

Chemical Identifiers

Chemical Name or Material Fumaramide
CAS 627-64-5
Molecular Formula C4H6N2O2
Formula Weight 114.11g/mol
MDL Number MFCD00025527
Beilstein 1721338
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2520014

 
alfa aesar™
B25200-14
25g Each for $64.15

AAB2520022

 
alfa aesar™
B25200-22
100g Each for $529.65
Description & Specifications

Specifications

Melting Point ≈266°C (dec.)
Quantity 25g
Chemical Name or Material Fumaramide
CAS 627-64-5
Assay 97%
Molecular Formula C4H6N2O2
Formula Weight 114.11g/mol
MDL Number MFCD00025527
Beilstein 1721338