missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaraldehyde Bis(dimethyl Acetal) 90.0+%, TCI America™
$70.84 - $205.05
Chemical Identifiers
| CAS | 6068-62-8 |
|---|---|
| Molecular Formula | C8H16O4 |
| Molecular Weight (g/mol) | 176.212 |
| MDL Number | MFCD00075507 |
| InChI Key | ZFGVCDSFRAMNMT-AATRIKPKSA-N |
| Synonym | fumaraldehyde bis dimethyl acetal, e-1,1,4,4-tetramethoxybut-2-ene, fumaraldehyde bis dimethylacetal, 2-butenedial bis dimethyl acetal, 1,1,4,4-tetramethoxy-trans-2-butene, trans-1,1,4,4-tetramethoxy-2-butene, 1,1,4,4-tetramethoxy-but-2-ene |
| PubChem CID | 5862719 |
| IUPAC Name | (E)-1,1,4,4-tetramethoxybut-2-ene |
| SMILES | COC(C=CC(OC)OC)OC |
Chemical Identifiers
| 6068-62-8 | |
| 176.212 | |
| ZFGVCDSFRAMNMT-AATRIKPKSA-N | |
| 5862719 | |
| COC(C=CC(OC)OC)OC |
| C8H16O4 | |
| MFCD00075507 | |
| fumaraldehyde bis dimethyl acetal, e-1,1,4,4-tetramethoxybut-2-ene, fumaraldehyde bis dimethylacetal, 2-butenedial bis dimethyl acetal, 1,1,4,4-tetramethoxy-trans-2-butene, trans-1,1,4,4-tetramethoxy-2-butene, 1,1,4,4-tetramethoxy-but-2-ene | |
| (E)-1,1,4,4-tetramethoxybut-2-ene |
Specifications
| 6068-62-8 | |
| 103°C | |
| MFCD00075507 | |
| fumaraldehyde bis dimethyl acetal, e-1,1,4,4-tetramethoxybut-2-ene, fumaraldehyde bis dimethylacetal, 2-butenedial bis dimethyl acetal, 1,1,4,4-tetramethoxy-trans-2-butene, trans-1,1,4,4-tetramethoxy-2-butene, 1,1,4,4-tetramethoxy-but-2-ene | |
| COC(C=CC(OC)OC)OC | |
| 176.212 | |
| 176.21 | |
| Liquid |
| Colorless | |
| C8H16O4 | |
| 1 g | |
| ZFGVCDSFRAMNMT-AATRIKPKSA-N | |
| (E)-1,1,4,4-tetramethoxybut-2-ene | |
| 5862719 | |
| ≥90.0% (GC) | |
| Fumaraldehyde Bis(dimethyl Acetal) |
Safety and Handling
TSCA : No