Please login to your online account to display your discounted pricing

Alfa Aesar™ Fumagillin, 95%

Chemical Identifiers

Chemical Name or Material Fumagillin
CAS 23110-15-8
Formula Weight 458.55
MDL Number MFCD03990453
Merck Index 14,4286
Synonym Fumidil B; TNP-470
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67045MCR

 
alfa aesar™
J67045MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 458.55
Melting Point 194° to 195°C
Chemical Name or Material Fumagillin
Physical Form Solid
Assay 95%
CAS 23110-15-8
Merck Index 14,4286
Synonym Fumidil B; TNP-470
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
MDL Number MFCD03990453
Health Hazard 1 Warning
Color Off-white to yellow
Quantity 1mg
Odor Odorless
Sensitivity Air Sensitive
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
RTECS Number HE1750000
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

-30° to -10°C