Learn More
Fullerene powder, sublimed, 99.9+% C{60}, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 42008MD
Description
The optical absorption properties of C60 match solar spectrum in a way that suggests that C60-based films could be useful for photovoltaic applications. Because of its high electronic affinity it is one of the most common electron acceptor used in donor/acceptor based solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fullerene powder | |
99685-96-8 | |
1.7 g/mL | |
C60 | |
MFCD00151408 | |
UN1325 | |
Insoluble in water. Soluble in organic solvents. | |
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41 | |
720.66 | |
CHEBI:33128 | |
Powder |
Sublimed | |
Black | |
500°C to 600°C (sublimation) | |
C60 | |
250 mg | |
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih | |
XMWRBQBLMFGWIX-UHFFFAOYSA-N | |
(C\{60}-I\{h})[5,6]fullerene | |
123591 | |
≥99.9% |
Chemical Identifiers
99685-96-8 | |
720.66 | |
XMWRBQBLMFGWIX-UHFFFAOYSA-N | |
123591 | |
(C\{60}-I\{h})[5,6]fullerene |
C60 | |
MFCD00151408 | |
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih | |
CHEBI:33128 | |
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41 |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
RTECSNumber : LS9252500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only