missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fullerene powder, 99% C{6}{0}, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 046311MD

 View more versions of this product

Catalog No. AA46311MD



Description

Description

It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Fullerene powder
Black
500°C to 600°C (sublimation)
C60
250 mg
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
XMWRBQBLMFGWIX-UHFFFAOYSA-N
(C\{60}-I\{h})[5,6]fullerene
123591
99%
99685-96-8
1.7 g/mL
C60
MFCD00282904
UN1325
Soluble in organic solvents. Insoluble in water.
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
720.66
CHEBI:33128
Powder

Chemical Identifiers

99685-96-8
720.66
XMWRBQBLMFGWIX-UHFFFAOYSA-N
123591
(C\{60}-I\{h})[5,6]fullerene
C60
MFCD00282904
fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
CHEBI:33128
C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : LS9252500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only