missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fullerene powder, 99% C60

Manufacturer:  Alfa Aesar™ 4631106

 View more versions of this product

Catalog No. AA4631106



Specifications

Specifications

Fullerene powder
500°C to 600°C (sublimation)
5g
C60
MFCD00282904
fullerene, fullerene, buckminsterfullerene, buckyball, fullerene, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
C12=C3C4=C5C6=C1C7=C8C9=C1C%10=C%11C(=C29)C3=C2C3=C4C4=C5C5=C9C6=C7C6=C7C8=C1C1=C8C%10=C%10C%11=C2C2=C3C3=C4C4=C5C5=C%11C%12=C(C6=C95)C7=C1C1=C%12C5=C%11C4=C3C3=C5C(=C81)C%10=C23
720.66
CHEBI:33128
99%
1.7g/mL
Black
99685-96-8
C60
UN1325
XMWRBQBLMFGWIX-UHFFFAOYSA-N
(C\{60}-I\{h})[5,6]fullerene
123591
Powder

Chemical Identifiers

99685-96-8
720.66
XMWRBQBLMFGWIX-UHFFFAOYSA-N
123591
(C\{60}-I\{h})[5,6]fullerene
C60
MFCD00282904
fullerene, fullerene, buckminsterfullerene, buckyball, fullerene, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
CHEBI:33128
C12=C3C4=C5C6=C1C7=C8C9=C1C%10=C%11C(=C29)C3=C2C3=C4C4=C5C5=C9C6=C7C6=C7C8=C1C1=C8C%10=C%10C%11=C2C2=C3C3=C4C4=C5C5=C%11C%12=C(C6=C95)C7=C1C1=C%12C5=C%11C4=C3C3=C5C(=C81)C%10=C23
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : LS9252500

SDS Documents