Learn More
Thermo Scientific Chemicals Fullerene, nanotube, multi-walled, as-produced cathode deposits, core and shell, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04288604
Description
Multi-walled carbon nanotube were used in the comparative study of oxidative treatment to carbon nonmaterial's. Used as a reinforcement in CNT/SiC nano composites. Also used as fiber optics transducers, fiber optic tips for micro bubble generation in liquids, battery cathodes and high strength composites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fullerene | |
308068-56-6 | |
Black | |
UN1325 | |
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon | |
VNWKTOKETHGBQD-UHFFFAOYSA-N | |
methane | |
297 | |
Powder |
Nanotube, multi-walled, as-produced cathode deposits, core and shell | |
CH4 | |
MFCD00133992 | |
2 g | |
Insoluble in water and organic solvents. | |
C | |
16.043 | |
CHEBI:16183 |
Chemical Identifiers
308068-56-6 | |
16.043 | |
VNWKTOKETHGBQD-UHFFFAOYSA-N | |
297 | |
methane |
CH4 | |
MFCD00133992 | |
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon | |
CHEBI:16183 | |
C |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO â Research Use Only