missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fullerene Extract, C60 (contains ca. 20% C70), TCI America™

$22.92 - $134.45

Chemical Identifiers

CAS 99685-96-8
Molecular Formula C60
Molecular Weight (g/mol) 720.66
MDL Number MFCD00151408
InChI Key XMWRBQBLMFGWIX-UHFFFAOYSA-N
Synonym fullerene, fullerene, buckminsterfullerene, buckyball, fullerene, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
PubChem CID 123591
ChEBI CHEBI:33128
IUPAC Name (C\{60}-I\{h})[5,6]fullerene
SMILES C12=C3C4=C5C6=C1C7=C8C9=C1C%10=C%11C(=C29)C3=C2C3=C4C4=C5C5=C9C6=C7C6=C7C8=C1C1=C8C%10=C%10C%11=C2C2=C3C3=C4C4=C5C5=C%11C%12=C(C6=C95)C7=C1C1=C%12C5=C%11C4=C3C3=C5C(=C81)C%10=C23
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B1642100MG View Documents TCI America
B1642100MG
100mg Each for $22.92
 
B16421G missing translation for 'viewDocuments' TCI America
B16421G
1g Each for $134.45
 
Specifications

Chemical Identifiers

99685-96-8
720.66
XMWRBQBLMFGWIX-UHFFFAOYSA-N
123591
(C\{60}-I\{h})[5,6]fullerene
C60
MFCD00151408
fullerene, fullerene, buckminsterfullerene, buckyball, fullerene, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
CHEBI:33128
C12=C3C4=C5C6=C1C7=C8C9=C1C%10=C%11C(=C29)C3=C2C3=C4C4=C5C5=C9C6=C7C6=C7C8=C1C1=C8C%10=C%10C%11=C2C2=C3C3=C4C4=C5C5=C%11C%12=C(C6=C95)C7=C1C1=C%12C5=C%11C4=C3C3=C5C(=C81)C%10=C23

Specifications

Undesignated
99685-96-8
MFCD00151408
XMWRBQBLMFGWIX-UHFFFAOYSA-N
(C\{60}-I\{h})[5,6]fullerene
123591
720.66
≥70.0% (HPLC)
100mg
C60
fullerene, fullerene, buckminsterfullerene, buckyball, fullerene, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
C12=C3C4=C5C6=C1C7=C8C9=C1C%10=C%11C(=C29)C3=C2C3=C4C4=C5C5=C9C6=C7C6=C7C8=C1C1=C8C%10=C%10C%11=C2C2=C3C3=C4C4=C5C5=C%11C%12=C(C6=C95)C7=C1C1=C%12C5=C%11C4=C3C3=C5C(=C81)C%10=C23
720.66
CHEBI:33128
Crystal-Powder at 20°C
Fullerene Extract, C60 (contains ca. 20% C70)
Safety and Handling

Safety and Handling

RTECSNumber : LS9252500

TSCA : No