missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fullerene, carbon nanotube, multi-walled, ≤8 nm OD, 2-5 nm ID, 0.5-2 micron long

Manufacturer:  Alfa Aesar™ 4479006

 View more versions of this product

Catalog No. AA4479006



Specifications

Specifications

Fullerene, carbon nanotube
Multi-walled, OD: ≤8nm, ID: 2 to 5nm, 0.5 to 2 micron long
CH4
UN1325
VNWKTOKETHGBQD-UHFFFAOYSA-N
methane
297
Powder
5g
308068-56-6
MFCD00151408
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
C
16.043
CHEBI:16183

Chemical Identifiers

308068-56-6
16.043
VNWKTOKETHGBQD-UHFFFAOYSA-N
297
methane
CH4
MFCD00151408
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
CHEBI:16183
C
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

SDS Documents