missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Fullerene, buckytube/nanotube, single walled, >60% SWNT

Manufacturer:  Alfa Aesar™ 4450102

 View more versions of this product

Catalog No. AA4450102



Specifications

Specifications

Fullerene
0.5g
308068-56-6
MFCD00282904
biogas
VNWKTOKETHGBQD-UHFFFAOYSA-N
methane
297
Powder
Black
Buckytube/nanotube, single walled, >60% SWNT
CH4
UN1325
Soluble in benzene,toluene and chloroform.
C
16.043
CHEBI:16183

Chemical Identifiers

308068-56-6
16.043
VNWKTOKETHGBQD-UHFFFAOYSA-N
297
methane
CH4
MFCD00282904
biogas
CHEBI:16183
C
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents