Please login to your online account to display your discounted pricing

Alfa Aesar™ Fullerene, buckytube/nanotube, multi-walled, ground core, 7-12nm OD, 0.5-10 micron long

Chemical Identifiers

Chemical Name or Material Fullerene, buckytube/nanotube
CAS 308068-56-6
Name Note Multi-walled, ground core, 7 to 12nm OD, 0.5 to 10 micron long
MDL Number MFCD00282904
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4154903

 
alfa aesar™
4154903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4154914

 
alfa aesar™
4154914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4154906

 
alfa aesar™
4154906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Fullerene, buckytube/nanotube
Name Note Multi-walled, ground core, 7 to 12nm OD, 0.5 to 10 micron long
Physical Form Powder
MDL Number MFCD00282904
CAS 308068-56-6
UN Number UN1325
Health Hazard 1 Warning
Color Black
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Mesh Size −100 Mesh
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.