missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Full-Sized Ripple Tank

Best device for demonstrating wave mechanics to your students.

Manufacturer:  Fisher Science Education 15415

 View more versions of this product

Catalog No. S52078-1


 

Requires : Light source and rippler motor power supply

Specifications Description & Specifications

Specifications

For demonstrating wave mechanics to students
Tank: 23.6 in.; Glass bottom: 17 in.
Ripple tank
Tank: 23.6 in.; Glass bottom: 17 in.
40cm motor driven rippler bar, a hand rippler bar, vertical light support, 4 paraffin blocks, 20x28cm glass plate, parabolic reflector, and white plastic viewing screen
Tank: 60cm; Glass bottom: 43cm
1.5V
Tank: 60cm; Glass bottom: 43cm