Please login to your online account to display your discounted pricing

Ftorafur, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  197700010

 View more versions of this product

Catalog No. AC197700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Very Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 200.17
Color White
Quantity 1g
EINECS Number 241-846-2
TSCA TSCA
Melting Point 168° to 171°C
RTECS Number YR0450000
Merck Index 15, 9252
Synonym 5-Fluoro-1-(tetrahydro-2-furyl)uracil
Chemical Name or Material Ftorafur
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9FN2O3
MDL Number MFCD00012351
CAS 17902-23-7

Specifications

Chemical Name or Material Ftorafur
Name Note 99%
CAS 17902-23-7
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H9FN2O3
Formula Weight 200.17
MDL Number MFCD00012351
Synonym 5-Fluoro-1-(tetrahydro-2-furyl)uracil