Please login to your online account to display your discounted pricing

Ftorafur, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ftorafur
Name Note 99%
CAS 17902-23-7
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H9FN2O3
Formula Weight 200.17
MDL Number MFCD00012351
Synonym 5-Fluoro-1-(tetrahydro-2-furyl)uracil
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC197702500

  SDS
thermo fisher scientific
197702500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC197700010

  SDS
thermo fisher scientific
197700010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Very Fine Crystalline Powder
Formula Weight 200.17
Color White
Quantity 250mg
Merck Index 15, 9252
EINECS Number 241-846-2
TSCA TSCA
Melting Point 168° to 171°C
RTECS Number YR0450000
Chemical Name or Material Ftorafur
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H9FN2O3
MDL Number MFCD00012351
CAS 17902-23-7
Synonym 5-Fluoro-1-(tetrahydro-2-furyl)uracil