Please login to your online account to display your discounted pricing

Fructose GenAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Fructose
Name Note Low Glucose
CAS 57-48-7
Molecular Formula C6H12O6
Formula Weight 180.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK775610

 
avantor performance
775665
STAKMOR 10kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.16
CAS 57-48-7
Packaging STAKMOR
Name Note Low Glucose
Molecular Formula C6H12O6
Chemical Name or Material Fructose
Quantity 10kg