missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fructose GenAR, Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance 775665

 View more versions of this product

Catalog No. MK775610

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Fructose
10kg
57-48-7
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:48095
STAKMOR
Low Glucose
C6H12O6
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
180.16

Chemical Identifiers

57-48-7
180.156
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O
C6H12O6
BJHIKXHVCXFQLS-UYFOZJQFSA-N
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Documents