Invitrogen™ FRK (PTK5) Recombinant Human Protein DFS Item
FRK (PTK5) is a nuclear protein.
Manufacturer:  Invitrogen™ LSPV4192
 View more versions of this product
Catalog No. PV4192
Description
Description
FRK (PTK5) may function during G1 and S phase of the cell cycle, suppressing growth.
Specifications
Specifications
NP_002022
Frozen
86.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
SRC Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
FRK (PTK5), GTK, RAK
Baculovirus
Human
GST Tag
Documents