Friedrich & Dimmock Borosilicate Glass Melting Point Capillaries
Manufacturer:  Friedrich & Dimmock 90MMX15
 View more versions of this product
Catalog No. 12-141-5
Description
Description
  • Thin-walled capillaries
  • O.D., 1.5 to 2.0mm
  • Packed 100 capillaries per vial
Specifications
Specifications
One end sealed
1.5 to 2 mm
Melting Point Capillary
Borosilicate Glass
0.059 to 0.078 in.
3.54 in.
90 mm