LaMotte™ Fresh Water Test Kit
Teaches sampling techniques, careful measurement and data recording.
Manufacturer: LaMotte™ LAMR5902
Includes:
- Six individual cases that house components needed for various chemical tests
- Sampling bottles
- Limnology handbook
- Analysis report forms
- 50 tests per factor, except nitrate (40) and pH (100)
Description
- Combines limnology with a water testing kit
- Tests for dissolved oxygen, carbon dioxide, nitrates, phosphates, silica, pH, calcium, magnesium and total hardness
- Soft-side tote is made of durable fabric and a solid plastic bottom ready for field work
- Handy side pockets hold thermometers, pens, writing tablets and other tools
Order Info
Grades 5+.
Specifications
5+ | |
Testing and study of freshwater systems such as ponds, lakes, wetlands, rivers, streams | |
Refill Unit | |
Six individual cases that house components needed for various chemical tests, sampling bottles, limnology handbook, analysis report forms, 50 tests per factor, except nitrate (40) and pH (100) |