United Scientific Supplies Fractional Brass Masses
Mass set meets weighing standards.
Manufacturer:  United Scientific Supplies 61014EMD
 View more versions of this product
Catalog No. S40264
Description
Description
Set comes in compact storage case and includes masses from 500mg to 1g.
  • Individual masses for calibration
  • Build your own customized set
  • Replace missing or damaged masses
Specifications
Specifications
Fractional weight
Nickel Plated Brass
10g
Weight, Compact storage case
Documents